Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 S protein, His Tag SPN-C52Hd is expressed from human 293 cells HEK293. It contains AA Val 16 - Pro 1213 F817P, A892P, A89 9P, A942P,K986P,V987P Accession QHD43416.1 F817P, A892P, A899P, A942P,K986P,V987P. Proline substitutions F817P, A89 P 1 * 50 µG
Catalog # ACRBSPN-C52HD-50UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein, His Tag SPN-C52Hd is expressed from human 293 cells HEK293. It contains AA Val 16 - Pro 1213 F817P, A892P, A89 9P, A942P,K986P,V987P Accession QHD43416.1 F817P, A892P, A899P, A942P,K986P,V987P. Proline substitutions F817P, A89 P 1 * 50 µG
Catalog # ACRBSPN-C52HD-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together