Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Cynomolgus NKp46, His,Avitag NC1-C82E5 is expressed from human 293 cells HEK293. It contains AA Pro 22 - Arg 257 Access ion Q95JB9-1. 1 * 25 µG
Catalog # ACRBNC1-C82E5-25UG
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus NKp46, His,Avitag NC1-C82E5 is expressed from human 293 cells HEK293. It contains AA Pro 22 - Arg 257 Access ion Q95JB9-1. 1 * 25 µG
Catalog # ACRBNC1-C82E5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together