Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human ACE2, His,Avitag AC2-H82E7 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Ser 740 Accession Q 9BYF1-1. 1 * 100 mg
Catalog # ACRBAC2-H82E7100MG
Supplier:  ACRO Biosystems
Biotinylated Human ACE2, His,Avitag AC2-H82E7 is expressed from human 293 cells HEK293. It contains AA Gln 18 - Ser 740 Accession Q 9BYF1-1. 1 * 100 mg
Catalog # ACRBAC2-H82E7100MG
Supplier:  ACRO Biosystems
Frequently Bought Together