Order Entry
Ireland
ContactUsLinkComponent
An antibody specific for GSTM3 has been pre-coated onto a 96-wellplate (12 x 8 Well Strips). Standards or test samples are added to the wells, incubated and removed. A biotinylated detector antibody specific for GSTM3 is added, incubated and followed 1 * 1 KIT
Catalog # AVIVOKCD01110
An antibody specific for GSTM3 has been pre-coated onto a 96-wellplate (12 x 8 Well Strips). Standards or test samples are added to the wells, incubated and removed. A biotinylated detector antibody specific for GSTM3 is added, incubated and followed 1 * 1 KIT
Catalog # AVIVOKCD01110
Frequently Bought Together