KLERCIDE LOW RESIDUE QUAT METERED DOSE CONCENTRATE Filtered 1 * 5 L
Catalog # SHIE6600074
Supplier: ECOLAB CONTAMINATION CONTROL
KLERCIDE LOW RESIDUE QUAT METERED DOSE CONCENTRATE Filtered 1 * 5 L
Catalog # SHIE6600074
Supplier: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Biocide, sterile, ready to use, Klercide™ Sporicidal Active Chlorine
Supplier: ECOLAB CONTAMINATION CONTROL
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Supplier: Dr. Weigert
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals