Order Entry
Ireland
ContactUsLinkComponent
Glove R299 High visibility cut resistant liner with Spandex back and durable palm with Kevlar stitching. Thermoplastic Rubber bumpers. Extended airprene wrist closure. Size 9 1 * 5 PAIR
Catalog # ANSER299090
Supplier:  Ansell
Glove R299 High visibility cut resistant liner with Spandex back and durable palm with Kevlar stitching. Thermoplastic Rubber bumpers. Extended airprene wrist closure. Size 9 1 * 5 PAIR
Catalog # ANSER299090
Supplier:  Ansell
Frequently Bought Together