Glove R299 High visibility cut resistant liner with Spandex back and durable palm with Kevlar® stitching. Thermoplastic Rubber bumpers. Extended airprene wrist closure. Size 8 1 * 5 PAIR
Catalog # ANSER299080
Supplier: Ansell
Frequently Bought Together