Order Entry
Ireland
ContactUsLinkComponent
Glove R176 Medium duty cut and sewn impact glove with 70g thinsulate and barrier for liquid resistance and cut and cold protection. Palm features synthetic leather with PVC patches for extra grip. Touch Screen compatible on first two fingers and thum 1 * 5 PAIR
Catalog # ANSER176120
Supplier:  Ansell
Glove R176 Medium duty cut and sewn impact glove with 70g thinsulate and barrier for liquid resistance and cut and cold protection. Palm features synthetic leather with PVC patches for extra grip. Touch Screen compatible on first two fingers and thum 1 * 5 PAIR
Catalog # ANSER176120
Supplier:  Ansell
Frequently Bought Together