Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
NOTE: THESE FILTERS ARE NOT FOR USE WITH OXYGEN. COMPATIBLE WITH ICP-MS INSTRUMENTS SUCH AS THE NEXION 1000/2000/350/300. 1 * 1 items
Catalog # PERCN8146005
Supplier:
PerkinElmer
Expand image
NOTE: THESE FILTERS ARE NOT FOR USE WITH OXYGEN. COMPATIBLE WITH ICP-MS INSTRUMENTS SUCH AS THE NEXION 1000/2000/350/300. 1 * 1 items
Catalog # PERCN8146005
Supplier:
PerkinElmer
Frequently Bought Together
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Accessories for Transferpette® electronic pipettes
Supplier: Brand
Fixing cover device 1 * 1 items
Supplier: VWR Collection
Spare charging adapter, for Eppendorf Xplorer®, electronic Multipettes®, and charger stands 1 * 1 items
Supplier: EPPENDORF
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: Brand
Filter,membrane for research pipette 1 * 50 items
Supplier: EPPENDORF
Accessories for accu-jet® pro
Supplier: Brand
Accessories for water purification millipore
Supplier: Merck
Temperature sensors for IKA digital dry block heaters
Supplier: IKA