Order Entry
Ireland
ContactUsLinkComponent
THE HYPER SKIMMER REMOVAL TOOL IS USED WITH THE NEXION 5000 ICP-MS INSTRUMENT. ALSO REQUIRES SAMPLER SKIMMER REMOVAL TOOL (N8160118) . 1 * 1 items
Catalog # PERCN8160119
Supplier:  PerkinElmer
THE HYPER SKIMMER REMOVAL TOOL IS USED WITH THE NEXION 5000 ICP-MS INSTRUMENT. ALSO REQUIRES SAMPLER SKIMMER REMOVAL TOOL (N8160118) . 1 * 1 items
Catalog # PERCN8160119
Supplier:  PerkinElmer
Frequently Bought Together