Order Entry
Ireland
ContactUsLinkComponent
FRIDGE, white exterior * refrigerant R600a * SOLID door RIGHT hinged * castors 360 swivel * e-sensor * 5 perforated wire shelf 430mm * 1 perforated wire shelf 300mm Including 5 years warranty on cabinets 1 * 1 items
  GRAM884500929
 :  GRAM
FRIDGE, white exterior * refrigerant R600a * SOLID door RIGHT hinged * castors 360 swivel * e-sensor * 5 perforated wire shelf 430mm * 1 perforated wire shelf 300mm Including 5 years warranty on cabinets 1 * 1 items
  GRAM884500929
 :  GRAM
Frequently Bought Together