Female Luer Thread Style to 200 Series Barb, 3/16 (4.8 mm) ID Tubing, Clear Polycarbonate 1 * 100 items
IBSAFTLL250-9
Female Luer Thread Style to 200 Series Barb, 3/16 (4.8 mm) ID Tubing, Clear Polycarbonate 1 * 100 items
IBSAFTLL250-9
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection