Sodium dihydrogen phosphate monohydrate EMPROVE EXPERT BP,USP 1 * 1 kg
Catalog # 1.37093.1000
Supplier: MERCK PRODUCTION CHEMICALS
Sodium dihydrogen phosphate monohydrate EMPROVE EXPERT BP,USP 1 * 1 kg
Catalog # 1.37093.1000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride low in endotoxins suitable for use as excipient EMPROVE exp Ph Eur,BP,JP,USP 1 * 1 kg
Supplier: MERCK PRODUCTION CHEMICALS
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
di-Sodium hydrogen phosphate dihydrate EMPROVE® bio Ph Eur,BP,USP This product cannot be returned after delivery 1 * 1 kg
Supplier: MERCK PRODUCTION CHEMICALS
Dichloromethane stabilised, EMPLURA®
Supplier: Merck
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals