IHILIC-P CLASSIC, HILIC COLUMN, PEEK-SS, 5um, 150x2.1mm, 200A 1 * 1 items
Catalog # HISM180.152.0520
Supplier: HICHROM
IHILIC-P CLASSIC, HILIC COLUMN, PEEK-SS, 5um, 150x2.1mm, 200A 1 * 1 items
Catalog # HISM180.152.0520
Supplier: HICHROM
Frequently Bought Together
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
WHEATON® Aluminium Seals, Unlined
Supplier: DWK Life Sciences
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
HPTLC plates silica gel 60, RP-modified layers, glass backed
Supplier: Merck
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection
Grounding cable, type 23
Supplier: S.C.A.T.