Order Entry
Ireland
ContactUsLinkComponent
SpectraMax® Glow Luminescence (LUM) (96) Detection Cartridge for 96-well plates, utilizes a unique, well-isolating design that signi ficantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax® PARADIGM® and/or Spect m 1 * 1 items
Catalog # MLDV0200-7014
Supplier:  Molecular Devices
SpectraMax® Glow Luminescence (LUM) (96) Detection Cartridge for 96-well plates, utilizes a unique, well-isolating design that signi ficantly reduces the level of cross talk from surrounding wells. Compatible with SpectraMax® PARADIGM® and/or Spect m 1 * 1 items
Catalog # MLDV0200-7014
Supplier:  Molecular Devices
Frequently Bought Together