Order Entry
Ireland
ContactUsLinkComponent
SpectraMax® Fluorescence Polarization (FP) (rhodamine) Detection Cartridge collects all the necessary data in a single read of a mic roplate. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. The resulting speed and performance du 1 * 1 items
Catalog # MLDV0200-7010
Supplier:  Molecular Devices
SpectraMax® Fluorescence Polarization (FP) (rhodamine) Detection Cartridge collects all the necessary data in a single read of a mic roplate. Compatible with SpectraMax® PARADIGM® and/or SpectraMax® i3 systems. The resulting speed and performance du 1 * 1 items
Catalog # MLDV0200-7010
Supplier:  Molecular Devices
Frequently Bought Together