Order Entry
Ireland
ContactUsLinkComponent
Soy Peptone is substituted for Tryptone. LB Lennox, a nutritionally rich medium, was developed by Lennox for the growth and maintenance of pure cultures of recombinant strains of E. coli. 1 * 500 g
  USBIL1505-01
Soy Peptone is substituted for Tryptone. LB Lennox, a nutritionally rich medium, was developed by Lennox for the growth and maintenance of pure cultures of recombinant strains of E. coli. 1 * 500 g
  USBIL1505-01
Frequently Bought Together