BIOPROCESS SPINNER FLASK, 250ML, DIMPLED BOTTOM INTERNAL IMPELLER, COMPLETE BIOPROCESS SPINNER FLASK, 250ML, DIMPLED BOTTOM INTERNAL IMPELLER, COMPLETE 1 * 250 mL
Catalog # GLPRCLS-1450-250
Supplier: GLASS PRECISION ENGINEERING
Frequently Bought Together