Order Entry
Ireland
ContactUsLinkComponent
SACCHARIMETER 0-30 0.1 BALLING TH 0-35C 1 * 1 items
Catalog # GERI70844
Supplier:  GERING
SACCHARIMETER 0-30 0.1 BALLING TH 0-35C 1 * 1 items
Catalog # GERI70844
Supplier:  GERING
Frequently Bought Together