Order Entry
Ireland
ContactUsLinkComponent
Mouse PCSK9, His Tag (PC9-M5228) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gln 694 (Accession AAH38085.1). 1 * 1 mg
Catalog # ACRBPC9-M5228-1MG
Supplier:  ACROBIOSYSTEMS
Mouse PCSK9, His Tag (PC9-M5228) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gln 694 (Accession AAH38085.1). 1 * 1 mg
Catalog # ACRBPC9-M5228-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together