Order Entry
Ireland
ContactUsLinkComponent
Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession NP000518.1). 1 * 1 mg
Catalog # ACRBLDR-H5224-1MG
Supplier:  ACROBIOSYSTEMS
Human LDL R, His Tag (LDR-H5224) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - Arg 788 (Accession NP000518.1). 1 * 1 mg
Catalog # ACRBLDR-H5224-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together