Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human IL-29 IFN-lambda 1 Protein, His Tag,Human IL-29, His Tag (IL9-H4228) is expressed from human 293 cells (HEK 293). It contains AA Met 1 - Thr 200 (Accession NP742152.1).This protein carries a polyhistidine tag at the C-terminu 1 * 50 µG
Catalog # ACRBIL9-H4228-50UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human IL-29 IFN-lambda 1 Protein, His Tag,Human IL-29, His Tag (IL9-H4228) is expressed from human 293 cells (HEK 293). It contains AA Met 1 - Thr 200 (Accession NP742152.1).This protein carries a polyhistidine tag at the C-terminu 1 * 50 µG
Catalog # ACRBIL9-H4228-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together