Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP00554 8617.1). 1 * 1 mg
Catalog # ACRBB72-C52HB-1MG
Supplier:  ACRO Biosystems
Cynomolgus B7-H2, His Tag (B72-C52Hb) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Thr 256 (Accession XP00554 8617.1). 1 * 1 mg
Catalog # ACRBB72-C52HB-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together