Order Entry
Ireland
ContactUsLinkComponent
Mouse Nectin-4, His Tag (NE4-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession Q8R007-1) . 1 * 1 mg
Catalog # ACRBNE4-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Mouse Nectin-4, His Tag (NE4-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 31 - Ser 347 (Accession Q8R007-1) . 1 * 1 mg
Catalog # ACRBNE4-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together