Order Entry
Ireland
ContactUsLinkComponent
Human CD24, Mouse IgG2a Fc Tag (CD4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25 063-1). 1 * 1 mg
Catalog # ACRBCD4-H5257-1MG
Supplier:  ACRO Biosystems
Human CD24, Mouse IgG2a Fc Tag (CD4-H5257) is expressed from human 293 cells (HEK293). It contains AA Ser 27 - Gly 59 (Accession P25 063-1). 1 * 1 mg
Catalog # ACRBCD4-H5257-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together