Order Entry
Ireland
ContactUsLinkComponent
Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). 1 * 1 mg
Catalog # ACRBLE3-H5228-1MG
Supplier:  ACROBIOSYSTEMS
Human CD4, His Tag (LE3-H5228) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Trp 390 (Accession AAH25782). 1 * 1 mg
Catalog # ACRBLE3-H5228-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together