Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBCD5-H5225-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human CD55 DAF Protein, His Tag,Human CD55, His Tag (CD5-H5225) is expressed from human 293 cells (HEK293). It co ntains AA Asp 35 - Ser 353 (Accession NP000565.1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBCD5-H5225-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together