Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human CD83 Protein, His Tag,Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Ala 143 (Accession AAH30830).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD3-H5223100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human CD83 Protein, His Tag,Human CD83, His Tag (CD3-H5223) is expressed from human 293 cells (HEK293). It contai ns AA Thr 20 - Ala 143 (Accession AAH30830).This protein carries a polyhistidine tag at the C-terminus. The protein h 1 * 100 µG
Catalog # ACRBCD3-H5223100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together