Order Entry
Ireland
ContactUsLinkComponent
Human CEACAM-6, His Tag (CE6-H5223) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession NP002474. 3). 1 * 100 µG
  ACRBCE6-H5223100UG
Human CEACAM-6, His Tag (CE6-H5223) is expressed from human 293 cells (HEK293). It contains AA Lys 35 - Gly 320 (Accession NP002474. 3). 1 * 100 µG
  ACRBCE6-H5223100UG
Frequently Bought Together