Order Entry
Ireland
ContactUsLinkComponent
Human CEACAM-8, His Tag (CE8-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession NP001807. 2). 1 * 100 µG
Catalog # ACRBCE8-H5224100UG
Supplier:  ACRO Biosystems
Human CEACAM-8, His Tag (CE8-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Ser 319 (Accession NP001807. 2). 1 * 100 µG
Catalog # ACRBCE8-H5224100UG
Supplier:  ACRO Biosystems
Frequently Bought Together