Order Entry
Ireland
ContactUsLinkComponent
Human C5a, Tag Free (C5A-H5116) is expressed from E.coli cells. It contains AA Leu 679 - Arg 751 (Accession P01031). 1 * 50 µG
Catalog # ACRBC5A-H5116-50UG
Supplier:  ACRO Biosystems
Human C5a, Tag Free (C5A-H5116) is expressed from E.coli cells. It contains AA Leu 679 - Arg 751 (Accession P01031). 1 * 50 µG
Catalog # ACRBC5A-H5116-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together