Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 1 mg
Catalog # ACRBTNB-H5255-1MG
Supplier:
ACROBIOSYSTEMS
Expand image
Human DcR3, Fc Tag (TNB-H5255) is expressed from human 293 cells (HEK293). It contains AA Val 30 - His 300 (Accession O95407-1). 1 * 1 mg
Catalog # ACRBTNB-H5255-1MG
Supplier:
ACROBIOSYSTEMS
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection