Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human Dkk-3 (R335G) Protein, His Tag,Human Dkk-3 (R335G), His Tag (DK3-H5220) is expressed from human 293 cells ( HEK293). It contains AA Ala 22 - Ile 350 (Accession Q9UBP4-1 (R335G)).This protein carries a polyhistidine tag at the 1 * 200 µG
Catalog # ACRBDK3-H5220200UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Dkk-3 (R335G) Protein, His Tag,Human Dkk-3 (R335G), His Tag (DK3-H5220) is expressed from human 293 cells ( HEK293). It contains AA Ala 22 - Ile 350 (Accession Q9UBP4-1 (R335G)).This protein carries a polyhistidine tag at the 1 * 200 µG
Catalog # ACRBDK3-H5220200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together