Order Entry
Ireland
ContactUsLinkComponent
ActiveMax® Mouse VEGF120, Tag Free (VE0-M4211) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accessio n AAB22254.1 ). 1 * 50 µG
Catalog # ACRBVE0-M4211-50UG
Supplier:  ACRO Biosystems
ActiveMax® Mouse VEGF120, Tag Free (VE0-M4211) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 146 (Accessio n AAB22254.1 ). 1 * 50 µG
Catalog # ACRBVE0-M4211-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together