Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human PD-L2, Fc,Avitag (PD2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on Q9BQ51-1). 1 * 25 µG
Catalog # ACRBPD2-H82F6-25UG
Supplier:  ACRO Biosystems
Biotinylated Human PD-L2, Fc,Avitag (PD2-H82F6) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accessi on Q9BQ51-1). 1 * 25 µG
Catalog # ACRBPD2-H82F6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together