Order Entry
Ireland
ContactUsLinkComponent
MABSol® Biotinylated Mouse PD-L1, Fc,Avitag (PD1-M82F5) is expressed from human HEK293 cells. It contains AA Phe 19 - Thr 238 (Acces sion NP068693). 1 * 200 µG
Catalog # ACRBPD1-M82F5200UG
Supplier:  ACROBIOSYSTEMS
MABSol® Biotinylated Mouse PD-L1, Fc,Avitag (PD1-M82F5) is expressed from human HEK293 cells. It contains AA Phe 19 - Thr 238 (Acces sion NP068693). 1 * 200 µG
Catalog # ACRBPD1-M82F5200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together