Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Mouse M-CSF R, Avitag,His Tag (CSR-M82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 ( Accession P09581-1). 1 * 200 µG
Catalog # ACRBCSR-M82E8200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse M-CSF R, Avitag,His Tag (CSR-M82E8) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Ser 511 ( Accession P09581-1). 1 * 200 µG
Catalog # ACRBCSR-M82E8200UG
Supplier:  ACRO Biosystems
Frequently Bought Together