Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human IL-21, Fc,Avitag (IL1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accessi on Q9HBE4-1). 1 * 200 µG
Catalog # ACRBIL1-H82F7200UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-21, Fc,Avitag (IL1-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Ser 155 (Accessi on Q9HBE4-1). 1 * 200 µG
Catalog # ACRBIL1-H82F7200UG
Supplier:  ACRO Biosystems
Frequently Bought Together