Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Human Akt1 Protein, His,Strep II Tag,Human Akt1, His,Strep II Tag (AK1-H5283) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Ala 480 (Accession AAH00479).This protein carries a polyhistidine tag at the C-termin 1 * 25 µG
Catalog # ACRBAK1-H5283-25UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Akt1 Protein, His,Strep II Tag,Human Akt1, His,Strep II Tag (AK1-H5283) is expressed from human 293 cells ( HEK293). It contains AA Met 1 - Ala 480 (Accession AAH00479).This protein carries a polyhistidine tag at the C-termin 1 * 25 µG
Catalog # ACRBAK1-H5283-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together