Recombinat Protein,Biotinylated Zika virus NS1 Protein, Avitag™,His Tag,Biotinylated Zika virus NS1, Avitag,His Tag (NS1-Z82E9) is e xpressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession ALU33341.1).This protein carries 1 * 25 µG
Catalog # ACRBNS1-Z82E9-25UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together