Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus CD16, His Tag (BLI verified) (FC6-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Acc ession Q8SPW2-1). 1 * 100 µG
Catalog # ACRBFC6-C52H9100UG
Supplier:  ACRO Biosystems
Cynomolgus CD16, His Tag (BLI verified) (FC6-C52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Acc ession Q8SPW2-1). 1 * 100 µG
Catalog # ACRBFC6-C52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together