Order Entry
Ireland
ContactUsLinkComponent
Mouse LAIR-1, Mouse IgG2a Fc Tag, low endotoxin (LA1-M5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Tyr 141 (Accession Q8BG84-1). 1 * 1 mg
Catalog # ACRBLA1-M5254-1MG
Supplier:  ACROBIOSYSTEMS
Mouse LAIR-1, Mouse IgG2a Fc Tag, low endotoxin (LA1-M5254) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Tyr 141 (Accession Q8BG84-1). 1 * 1 mg
Catalog # ACRBLA1-M5254-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together