Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human DNAM-1, Fc,Avitag (DN1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Access ion NP006557). 1 * 25 µG
Catalog # ACRBDN1-H82F9-25UG
Supplier:  ACRO Biosystems
Biotinylated Human DNAM-1, Fc,Avitag (DN1-H82F9) is expressed from human 293 cells (HEK293). It contains AA Glu 19 - Asn 247 (Access ion NP006557). 1 * 25 µG
Catalog # ACRBDN1-H82F9-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together