Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Mouse VEGF164, His,Avitag (VE4-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Acce ssion Q00731-2). 1 * 25 µG
Catalog # ACRBVE4-M82Q3-25UG
Supplier:  ACRO Biosystems
Biotinylated Mouse VEGF164, His,Avitag (VE4-M82Q3) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 190 (Acce ssion Q00731-2). 1 * 25 µG
Catalog # ACRBVE4-M82Q3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together