Order Entry
Ireland
ContactUsLinkComponent
Mouse B7-1, His Tag (CD0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 38 - Lys 245 (Accession Q00609-1). 1 * 1 mg
Catalog # ACRBCD0-M5228-1MG
Supplier:  ACROBIOSYSTEMS
Mouse B7-1, His Tag (CD0-M5228) is expressed from human 293 cells (HEK293). It contains AA Val 38 - Lys 245 (Accession Q00609-1). 1 * 1 mg
Catalog # ACRBCD0-M5228-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together