Order Entry
Ireland
ContactUsLinkComponent
Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession P35052- 1). 1 * 100 µG
Catalog # ACRBGP1-H52H9100UG
Supplier:  ACRO Biosystems
Human Glypican 1, His Tag (GP1-H52H9) is expressed from human 293 cells (HEK293). It contains AA Asp 24 - Ser 530 (Accession P35052- 1). 1 * 100 µG
Catalog # ACRBGP1-H52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together