Order Entry
Ireland
ContactUsLinkComponent
Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 1 mg
Catalog # ACRBTN1-H5222-1MG
Supplier:  ACROBIOSYSTEMS
Human TNFR1, His Tag (TN1-H5222) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Thr 211 (Accession NP001056). 1 * 1 mg
Catalog # ACRBTN1-H5222-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together