Order Entry
Ireland
ContactUsLinkComponent
Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession AA I13679). 1 * 100 µG
Catalog # ACRBPD2-H52A5100UG
Supplier:  ACRO Biosystems
Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession AA I13679). 1 * 100 µG
Catalog # ACRBPD2-H52A5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together