Order Entry
Ireland
ContactUsLinkComponent
Recombinat Protein,Mouse IL-4 R alpha CD124 Protein, Fc Tag,Mouse IL-4 R alpha, Fc Tag (ILR-M5252) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Arg 233 (Accession NP001008700).This protein carries a human IgG1 Fc tag at the C- 1 * 1 mg
Catalog # ACRBILR-M5252-1MG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Mouse IL-4 R alpha CD124 Protein, Fc Tag,Mouse IL-4 R alpha, Fc Tag (ILR-M5252) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Arg 233 (Accession NP001008700).This protein carries a human IgG1 Fc tag at the C- 1 * 1 mg
Catalog # ACRBILR-M5252-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together