Order Entry
Ireland
ContactUsLinkComponent
Human TNFSF11, Fc tag (RAL-H5265) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession AAC51762.1) . 1 * 1 mg
Catalog # ACRBRAL-H5265-1MG
Supplier:  ACROBIOSYSTEMS
Human TNFSF11, Fc tag (RAL-H5265) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession AAC51762.1) . 1 * 1 mg
Catalog # ACRBRAL-H5265-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together